Learn More
Aviva Systems Biology Corp GCDH Antibody - N-terminal region (ARP43568_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43568_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GCDH. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GCDH. Alternate names: ACAD5, GCDApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58571