Learn More
Eurofins Discoverx PathHunter GCGR Total Internalization HEK 293 Cell Line, 2 vials. (93-0820C1)
Catalog No. 502001351
Encompass
The PathHunter GCGR Total Internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line uses EFC technology to measure ligand-induced internalization of GCGR via detection of localization to the early endosome. The included cell line overexpresses EA-tagged GCGR and a ProLink-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the GCGR receptor to the early endosome leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1351 Supplier Eurofins Discoverx Supplier No. 930820C1
Add to Cart
Add to Cart
Request a Quote
The PathHunter GCGR Total Internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line uses EFC technology to measure ligand-induced internalization of GCGR via detection of localization to the early endosome. The included cell line overexpresses EA-tagged GCGR and a ProLink-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the GCGR receptor to the early endosome leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.