Learn More
Aviva Systems Biology Corp GCHFR Antibody - N-terminal region (ARP43574_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43574_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GCHFR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GCHFR. Alternate names: GFRP, HsT16933, MGC138467, MGC138469, P35ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58575