Learn More
Eurofins Discoverx GCK, active, 250 µg. (14-743M)
Supplier:  Eurofins Discoverx 14743M
Encompass
N-terminal GST-tagged recombinant, human GCK amino acids 1-473. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9274