Learn More
Aviva Systems Biology Corp GCKR Antibody - C-terminal region (ARP63697_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63697_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GCKR. It was validated on Western Blot. . Alternate names: GKRP, FGQTL5ApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-288-055