Learn More
Aviva Systems Biology Corp Gcm1 Antibody - middle region (ARP36862_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36862_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Gcm1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of mouse Gcm1. Alternate names: GCMa, Gcm-rs2, Gcm1-rs1, glideApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-284-528