Learn More
Cayman Chemical GCMS DrUG Stndard MIXure 1 1ea
SDP
Catalog No. 502895139
Encompass_Preferred
A mixture of common alkaloids and opioids, as well as a prominent metabolite; designed to provide authentic standards for analyses using GC- or LC-MS
Catalog No. 50-289-5139 Supplier Cayman Chemical Supplier No. 90027701 EA
A mixture of common alkaloids and opioids, as well as a prominent metabolite; designed to provide authentic standards for analyses using GC- or LC-MS