Learn More
Eurofins Discoverx GCN2, active, 10 µg. (14-934)
Supplier:  Eurofins Discoverx 14934
Encompass
N-terminal GST-tagged recombinant, human GCN2 full length. The recombinant protein contains the amino acid substitution E556G with reference to GenPept NP_001013725. E556G is reported in GenPept CAH10626. GCN2 (EIF2AK4) belongs to a family of kinases that phosphorylate the α subunit of eukaryotic translation initiation factor-2 (EIF2S1) to downregulate protein synthesis in response to varied cellular stresses. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9441