Learn More
Medchemexpress LLC HY-100877 10mg Medchemexpress, GCN2-IN-1 CAS:1448693-69-3 Purity:>98%
SDP
Catalog No. 501931983
Encompass_Preferred
Medchemexpress, HY-100877 10mg GCN2-IN-1 CAS:1448693-69-3 GCN2-IN-1 (A-92) is a potent general control nonderepressible 2 kinase ( GCN2 ) inhibitor with an IC 50 of <0.3 μM in the enzyme assay and an IC 50 of 0.3-3 μM in the cell assay [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1983 Supplier Medchemexpress LLC Supplier No. HY10087710MG
Medchemexpress, HY-100877 10mg GCN2-IN-1 CAS:1448693-69-3 GCN2-IN-1 (A-92) is a potent general control nonderepressible 2 kinase ( GCN2 ) inhibitor with an IC 50 of <0.3 μM in the enzyme assay and an IC 50 of 0.3-3 μM in the cell assay [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.