Learn More
Medchemexpress LLC HY-130240 5mg Medchemexpress, GCN2-IN-6 CAS:2183470-09-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1302405MG
Encompass_Preferred
Medchemexpress, HY-130240 5mg GCN2-IN-6 CAS:2183470-09-7 GCN2-IN-6 (Compound 6d) is a potent, and orally available GCN2 inhibitor confirmed by in-house enzymatic (IC50 of 1.8 nM) and cellular assays (IC50 of 9.3 nM). GCN2-IN-6 is also a eIF2α kinase PERK inhibitor with an IC50 of 0.26 nM (in enzymatic assay) and 230 nM (in cells). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0197