Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gcn2-in-6 (compound 6d) | 2183470-09-7 | 98.9% | C19H12Cl2F2N4O3S | 10MG
SDP

Catalog No. 5000260012
Encompass_Preferred

GCN2-IN-6 is a small-molecule inhibitor of the eIF2α kinase GCN2 used as a research tool to probe the integrated stress response and related signaling pathways. It has been characterized in enzymatic and cellular assays and is provided for laboratory research in solid and DMSO solution formats.

  • Potent GCN2 inhibitor with enzymatic IC50 ~1.8 nM and cellular IC50 ~9.3 nM.
  • Also shows activity against PERK in enzymatic and cellular assays.
  • High nominal purity (~98.9% as listed on product page).
  • Molecular formula C19H12Cl2F2N4O3S; molecular weight 485.29.
  • Available as mg-scale solid and 10 mM DMSO solutions for convenient use.
  • CAS 2183470-09-7 for unambiguous chemical identification.

Catalog No. 50-002-60012 Supplier Medchemexpress LLC Supplier No. HY13024010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GCN2-IN-6 is a small-molecule inhibitor of the eIF2α kinase GCN2 used as a research tool to probe the integrated stress response and related signaling pathways. It has been characterized in enzymatic and cellular assays and is provided for laboratory research in solid and DMSO solution formats.

  • Potent GCN2 inhibitor with enzymatic IC50 ~1.8 nM and cellular IC50 ~9.3 nM.
  • Also shows activity against PERK in enzymatic and cellular assays.
  • High nominal purity (~98.9% as listed on product page).
  • Molecular formula C19H12Cl2F2N4O3S; molecular weight 485.29.
  • Available as mg-scale solid and 10 mM DMSO solutions for convenient use.
  • CAS 2183470-09-7 for unambiguous chemical identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.