Learn More
Medchemexpress LLC HY-112654 100mg , GCN2iB CAS:2183470-12-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY112654100MG
Encompass_Preferred
HY-112654 100mg GCN2iB CAS: 2183470-12-2 GCN2iB is an ATP-competitive inhibitor of a serine/threonine-protein kinase general control nonderepressible 2 (GCN2), with an IC50 of 2.4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0554