Learn More
Medchemexpress LLC HY-112654 10mg Medchemexpress, GCN2iB CAS:2183470-12-2 Purity:>98%
SDP
Catalog No. 502028826
Encompass_Preferred
Medchemexpress, HY-112654 10mg GCN2iB CAS:2183470-12-2 GCN2iB is an ATP-competitive inhibitor of a serine/threonine-protein kinase general control nonderepressible 2 (GCN2), with an IC50 of 2.4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8826 Supplier Medchemexpress LLC Supplier No. HY11265410MG
Medchemexpress, HY-112654 10mg GCN2iB CAS:2183470-12-2 GCN2iB is an ATP-competitive inhibitor of a serine/threonine-protein kinase general control nonderepressible 2 (GCN2), with an IC50 of 2.4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.