Learn More
Selleck Chemical LLC GCN2iB 5mg
SDP
Catalog No. 502173439
Encompass_Preferred
GCN2iB is an ATP-competitive serine/threonine-protein kinase general control nonderepressible 2 (GCN2) inhibitor with IC50 of 2.4 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3439 Supplier Selleck Chemical LLC Supplier No. S89295MG
GCN2iB is an ATP-competitive serine/threonine-protein kinase general control nonderepressible 2 (GCN2) inhibitor with IC50 of 2.4 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.