Learn More
TARGETMOL CHEMICALS INC GCN2IB 5MG
SDP
Catalog No. 502581535
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GCN2iB is an ATP-competitive inhibitor of general control nonderepressible 2 (GCN2) with an IC50 of 2.4 nM. purity: 98%
Catalog No. 50-258-1535 Supplier TARGETMOL CHEMICALS INC Supplier No. T113755MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GCN2iB is an ATP-competitive inhibitor of general control nonderepressible 2 (GCN2) with an IC50 of 2.4 nM. purity: 98%