Learn More
Enzo Life Sciences GCN5 (human), (recombinant) (50 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE2720050
Encompass_Preferred
Alternative name: GCN5L2, Histone acetyltransferase GCN5. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 20mM sodium citrate, pH 6.0, containing 0.25M NaCl, 1mg/ml BSA and 10% glycerol. Source: Produced in E. coli. Catalytic domain (aa 497-663) of human hGCN5L. Long Term Storage: -80°C.
Catalog No. 50-201-1966