Learn More
Aviva Systems Biology Corp GCNT3 Antibody - C-terminal region (ARP43526_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43526_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GCNT3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human GCNT3. Alternate names: C2/4GnT, C2GnT-M, C2GnT2, GnT-M, GNTM, C24GNT, C2GNT2, C2GNTMApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Yeast
Catalog No. 50-014-53368