Learn More
Aviva Systems Biology Corp GCNT4 Antibody - C-terminal region (ARP47278_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47278_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GCNT4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human GCNT4. Alternate names: C2GNT3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59505