Learn More
Aviva Systems Biology Corp Gcom1 Antibody - middle region (ARP51834_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51834_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Gcom1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human Gcom1. Alternate names: FLJ30973, GRINL1A, Gcom2, Gup1, Gup2, MGC126694, MGC138353, gcom, MYZAP-POLR2MApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-60626