Learn More
Bio Rad Laboratories GCR96 CARTRIDGES PACK OF 10
Catalog No. NC1879382
Encompass
  • Designed for seamless operation with QX ONE ddPCR System.
  • Combines droplet generation, thermal cycling, and droplet reading in a single unit.
  • Enables walk-away operation for efficient sample processing.
  • Supports high-throughput analysis of up to 96 samples at once.
  • Pack includes 10 cartridges.
  • Ideal for ddPCR workflows.
Catalog No. NC1879382 Supplier Bio Rad Laboratories Supplier No. 12006859
  • Designed for seamless operation with QX ONE ddPCR System.
  • Combines droplet generation, thermal cycling, and droplet reading in a single unit.
  • Enables walk-away operation for efficient sample processing.
  • Supports high-throughput analysis of up to 96 samples at once.
  • Pack includes 10 cartridges.
  • Ideal for ddPCR workflows.