Learn More
Bio Rad Laboratories GCR96 FOIL HEAT SEAL PACK 100
Catalog No. NC1879383
Encompass
  • Durable and pierceable seal
  • Compatible with GCR96 cartridges
  • Designed for PCR applications
  • Specifically certified for QX ONE droplet digital PCR systems
  • Validated for use with PX1 PCR plate sealer
  • Pack quantity: 100
Catalog No. NC1879383 Supplier Bio Rad Laboratories Supplier No. 12006843
  • Durable and pierceable seal
  • Compatible with GCR96 cartridges
  • Designed for PCR applications
  • Specifically certified for QX ONE droplet digital PCR systems
  • Validated for use with PX1 PCR plate sealer
  • Pack quantity: 100