Learn More
Aviva Systems Biology Corp GCS1 Antibody - N-terminal region (ARP46708_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46708_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GCS1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GCS1. Alternate names: GCS1, CDG2BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53486