Learn More
ACROBiosystems Recombinant Protein;Human G-CSF R / CD114 Protein;HEK293;1MG;GCR,CSF3R,CD114,GCSFR,GCSF-R,G-CSF-R,CSF-3R
Catalog No. 501657609
Encompass_Preferred
Recombinant Protein;Human G-CSF R / CD114 Protein;1MG;Human G-CSF R, His Tag (GCR-H5223) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Pro 621 (Accession # NP_000751.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 69 kDa. The protein migrates as 94 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7609 Supplier ACROBiosystems Supplier No. GCRH52231MG
Recombinant Protein;Human G-CSF R / CD114 Protein;1MG;Human G-CSF R, His Tag (GCR-H5223) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Pro 621 (Accession # NP_000751.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 69 kDa. The protein migrates as 94 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.