Learn More
Aviva Systems Biology Corp GCSH Antibody - C-terminal region (ARP64203_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64203_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GCSH. It was validated on Western Blot. . Alternate names: GCE, NKHApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-288-203