Learn More
Sino Biological GCSH Antibody, Rabbit PAb 100uL
Supplier:  Sino Biological 14568RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human GCSH (rh GCSH; Catalog#14568-H07E; AAA36011.1; Ser49-Glu173). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-224-7974