Learn More
ATCC GCT; Lung Histiocytoma; Human (Homo sapiens)
Supplier:  ATCC TIB223
Encompass
GCT [Giant Cell Tumor] was isolated from the lung of a 29-year-old male patient with fibrous histiocytoma. The cell line produces CSA for human granulocyte precursors and EEA for erythroid precursor. Medium conditioned by this line can be used as a source of prostaglandin E and plasminogen activator.
Catalog No. 50-238-2783
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.