Learn More
Waters Corp Waters Corporation GCT Premier Standards Kit, EI, CI, DCI, SIP
Catalog No. 50201674 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Mass Spectrometry (GCT Premier, EI, CI, DCI, SIP) standards kit for system assessment.
  • Comprehensive kit includes 12 components for various assessment needs.
  • Includes standards for resolution specification, tuning, and calibration.
  • Contains standards for GC/MS, DIP EI +ve, DIP CI +ve, GC–EI +ve, GC–CI +ve, GC–CI –ve, DIP CI –ve, and DCI +ve mass measurement accuracy and sensitivity specifications.
  • Features hexachlorobenzene for DRE demonstration.
  • Supplied with glass vials, Pasteur pipettes, and a latex pipette bulb.
  • A certificate of analysis is provided.
  • Designed for room temperature storage and ambient shipping.
  • Has a shelf life of 6 months.
  • Ideal for system performance applications.
Catalog No. 50-201-674 Supplier Waters Corp Supplier No. 700002878
Mass Spectrometry (GCT Premier, EI, CI, DCI, SIP) standards kit for system assessment.
  • Comprehensive kit includes 12 components for various assessment needs.
  • Includes standards for resolution specification, tuning, and calibration.
  • Contains standards for GC/MS, DIP EI +ve, DIP CI +ve, GC–EI +ve, GC–CI +ve, GC–CI –ve, DIP CI –ve, and DCI +ve mass measurement accuracy and sensitivity specifications.
  • Features hexachlorobenzene for DRE demonstration.
  • Supplied with glass vials, Pasteur pipettes, and a latex pipette bulb.
  • A certificate of analysis is provided.
  • Designed for room temperature storage and ambient shipping.
  • Has a shelf life of 6 months.
  • Ideal for system performance applications.