Learn More
Waters Corp Waters Corporation GCT Premier (FI, FD) Standards Kit, MS, Liquid
Supplier: Waters Corp 700002881
This standards kit is designed for system assessment in Mass Spectrometry (GCT Premier, FI, FD) applications. It includes six components for resolution, tuning, calibration, and sensitivity, along with glass vials, pipets, a latex bulb, and a Certificate of Analysis.
- Separation mode: GC
- Detection method: MS
- Formulation: Liquid
- Storage condition: Room temperature
- System type: MS
- Instrument: GC
- Shelf life: 6 months
- Shipping condition: Ambient
- Type: Reference
- Application: System performance
- Product type: Standards
- Units per package: 1 pk
- Hazardous materials requiring special shipping
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.