Learn More
Enzo Life Sciences UNC 45a monoclonal antibody (AbS1) (200 µg)
SDP
Supplier:  Enzo Life Sciences ADISRA1800F
Encompass_Preferred
Clone: AbS1. Host: Mouse. Isotype: IgG2b. Immunogen: Recombinant human UNC45. UniProt: Q9H3U1. Source: Purified from mouse ascites. Species Reactivity: Human, Mouse, Rat. Applications: WB. Recommended dilutions: Western Blot (1:1,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-810