Learn More
IBL America GD-IGA1 (Galactose-Deficient IGA1) ELISA
Supplier:  IBL America 27600
Encompass_Preferred
Solid phase sandwich ELISA for the determination of Gd-IgA1 (Galactose-deficient IgA1) in human serum and EDTA-plasma. For research use only, not for use in diagnostic procedures.As a primary antibody is coated on a plate, samples and standard are added intothe wells for 1st reaction. After the reaction, HRP-conjugated secondary antibody isadded into the wells for 2nd reaction. After washing away unbound the secondaryantibody, Tetra Methyl Benzidine (TMB) is added to the wells and color develops.
Catalog No. 50-136-6585