Learn More
Enzo Life Sciences Ganglioside GD1a. disodium salt (bovine brain) (1mg). CAS: 12707-58-3
SDP
Supplier:  Enzo Life Sciences ALX302007M001
Encompass_Preferred
Differentiation marker for cell growth. Protein kinase C (PKC) inhibitor. The major ganglioside of the nervous system. Alternative name: Disialoganglioside GD1a . 2Na (bovine brain), GD1a . 2Na. Purity: ≥98% (TLC). Appearance: Lyophilized. Solubility: Soluble in chloroform/methanol/DI water, 2:1:0.1. Long Term Storage: -20°C.
Catalog No. 50-200-8646