Learn More
Enzo Life Sciences Ganglioside GD1a. disodium salt (bovine brain) (5mg). CAS: 12707-58-3
SDP
Catalog No. 502008647
Encompass_Preferred
Differentiation marker for cell growth. Protein kinase C (PKC) inhibitor. The major ganglioside of the nervous system. Alternative name: Disialoganglioside GD1a . 2Na (bovine brain), GD1a . 2Na. Purity: ≥98% (TLC). Solubility: Soluble in chloroform/methanol/DI water, 2:1:0.1. Long Term Storage: -20°C.
Catalog No. 50-200-8647 Supplier Enzo Life Sciences Supplier No. ALX302007M005
Add to Cart
Add to Cart
Differentiation marker for cell growth. Protein kinase C (PKC) inhibitor. The major ganglioside of the nervous system. Alternative name: Disialoganglioside GD1a . 2Na (bovine brain), GD1a . 2Na. Purity: ≥98% (TLC). Solubility: Soluble in chloroform/methanol/DI water, 2:1:0.1. Long Term Storage: -20°C.