Learn More
Enzo Life Sciences Ganglioside GD3. disodium salt (bovine buttermilk) (1mg). CAS: 62010-37-1
SDP
Catalog No. 502008649
Encompass_Preferred
Induces mitochondrial permeability transition (MPT) without requiring elevated Ca2+ levels and thus triggers Fas-mediated apoptosis. Alternative name: Disialoganglioside GD3 . 2Na (bovine buttermilk), GD3 . 2Na (bovine buttermilk). Purity: ≥98% (TLC). Appearance: Lyophilized. Solubility: Soluble in chloroform/methanol, 2:1. Long Term Storage: -20°C.
Catalog No. 50-200-8649 Supplier Enzo Life Sciences Supplier No. ALX302010M001
Induces mitochondrial permeability transition (MPT) without requiring elevated Ca2+ levels and thus triggers Fas-mediated apoptosis. Alternative name: Disialoganglioside GD3 . 2Na (bovine buttermilk), GD3 . 2Na (bovine buttermilk). Purity: ≥98% (TLC). Appearance: Lyophilized. Solubility: Soluble in chloroform/methanol, 2:1. Long Term Storage: -20°C.