Learn More
Aviva Systems Biology Corp GDA Antibody - middle region (ARP63054_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63054_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GDA. It was validated on Western Blot. . Alternate names: CYPIN, GUANASE, KIAA1258, MGC9982, NEDASINApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-856