Learn More
Aviva Systems Biology Corp GDAP1L1 Antibody - N-terminal region (ARP49791_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49791_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GDAP1L1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GDAP1L1. Alternate names: DKFZp761K228, dJ881L22.1, dJ995J12.1.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60185