Learn More
Aviva Systems Biology Corp GDAP2 Antibody - N-terminal region (ARP57005_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57005_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GDAP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GDAP2. Alternate names: FLJ20142, dJ776P7.1, MACROD3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62849