Learn More
Apexbio Technology LLC GDC-0032, 10mg. Cas: 1282512-48-4 MFCD: MFCD26142641. PI3Kα inhibitor
SDP
Catalog No. 501905201
Encompass_Preferred
GDC-0032 is a novel and potent benzoxepin inhibitor of PI3K isoform with the Ki value of 0.29nM for PI3K[1].GDC-0032 has been reported to potently inhibit PI3K and have the selectivity over other PI3Ks. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5201 Supplier Apexbio Technology LLC Supplier No. B104710
Add to Cart
Add to Cart
GDC-0032 is a novel and potent benzoxepin inhibitor of PI3K isoform with the Ki value of 0.29nM for PI3K[1].GDC-0032 has been reported to potently inhibit PI3K and have the selectivity over other PI3Ks. Other sizes are available. Please inqury us for quote.