Learn More
Medchemexpress LLC HY-15186 100mg , GDC-0068 Ipatasertib;RG7440 CAS:1001264-89-6 Purity:98%
SDP
Catalog No. 501492723
Encompass_Preferred
Medchemexpress, HY-15186 100mg GDC-0068 Ipatasertib;RG7440 CAS:1001264-89-6 Formula:C24H32ClN5O2 IC50: 5±7 nM (Akt1), 18±10 nM (Akt2), 8±9 nM (Akt3), 3100±705 nM (PKA) Purity:98% GDC-0068 is a highly selective pan-Akt inhibitor targeting Akt1 / Akt2 / Akt3 with IC 50 of 5/18/8 nM, 620-fold selectivity over PKA. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2723 Supplier Medchemexpress LLC Supplier No. HY15186100MG
Medchemexpress, HY-15186 100mg GDC-0068 Ipatasertib;RG7440 CAS:1001264-89-6 Formula:C24H32ClN5O2 IC50: 5±7 nM (Akt1), 18±10 nM (Akt2), 8±9 nM (Akt3), 3100±705 nM (PKA) Purity:98% GDC-0068 is a highly selective pan-Akt inhibitor targeting Akt1 / Akt2 / Akt3 with IC 50 of 5/18/8 nM, 620-fold selectivity over PKA. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.