Learn More
Selleck Chemical LLC Ipatasertib (GDC-0068) 5mg 1001264-89-6 RG7440
SDP
Catalog No. NC0671901
Encompass_Preferred
Ipatasertib (GDC-0068, RG7440) is a highly selective pan-Akt inhibitor targeting Akt1/2/3 with IC50 of 5 nM/18 nM/8 nM in cell-free assays, 620-fold selectivity over PKA. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0671901 Supplier Selleck Chemical LLC Supplier No. S28085MG
Ipatasertib (GDC-0068, RG7440) is a highly selective pan-Akt inhibitor targeting Akt1/2/3 with IC50 of 5 nM/18 nM/8 nM in cell-free assays, 620-fold selectivity over PKA. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.