Learn More
Apexbio Technology LLC GDC-0068 (RG7440), 50mg. Cas: 1001264-89-6 MFCD: MFCD22124514. Pan-AKT inhibitor,highly selective
SDP
Supplier:  Apexbio Technology LLC A300650
Encompass_Preferred
GDC-0068 (RG7440) is a highly selective small molecule inhibitor of Akt with IC50s of 5, 18 and 8 nM for Akt1, Akt2 and Akt3. The PI3K-AKT pathway regulates cell growth, survival and tumorigenesis. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4777