Learn More
Medchemexpress LLC HY-101562 10mg Medchemexpress, GDC-0077 CAS:2060571-02-8 Purity:>98%
SDP
Catalog No. 502028666
Encompass_Preferred
Medchemexpress, HY-101562 10mg GDC-0077 CAS:2060571-02-8 GDC-0077 (RG6114) is a potent, orally available, and selective PI3Kα inhibitor (IC50=0.038 nM). GDC-0077 (RG6114) exerts its activity by binding to the ATP binding site of PI3K, thereby inhibiting the phosphorylation of PIP2 to PIP3. GDC-0077 (RG6114) is more selective for mutant versus wild-type PI3Kα. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8666 Supplier Medchemexpress LLC Supplier No. HY10156210MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-101562 10mg GDC-0077 CAS:2060571-02-8 GDC-0077 (RG6114) is a potent, orally available, and selective PI3Kα inhibitor (IC50=0.038 nM). GDC-0077 (RG6114) exerts its activity by binding to the ATP binding site of PI3K, thereby inhibiting the phosphorylation of PIP2 to PIP3. GDC-0077 (RG6114) is more selective for mutant versus wild-type PI3Kα. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.