Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GDC-0276 10mg | 1494581-70-2 | 10 MG
SDP

Catalog No. 5000226619
Encompass_Preferred

GDC-0276 is a small-molecule NaV1.7 (voltage-gated sodium channel 1.7) inhibitor supplied as a solid white to off-white powder for research use only. It is provided with supporting documentation including a datasheet, COA, and SDS.

  • High purity 99.69%
  • Molecular formula C24H31FN2O4S
  • Molecular weight 462.58
  • Physical form solid, white to off-white
  • Supplied as a 10 mg vial
  • For research use only; not for human or diagnostic use

Catalog No. 50-002-26619 Supplier Medchemexpress LLC Supplier No. HY11423710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

GDC-0276 is a small-molecule NaV1.7 (voltage-gated sodium channel 1.7) inhibitor supplied as a solid white to off-white powder for research use only. It is provided with supporting documentation including a datasheet, COA, and SDS.

  • High purity 99.69%
  • Molecular formula C24H31FN2O4S
  • Molecular weight 462.58
  • Physical form solid, white to off-white
  • Supplied as a 10 mg vial
  • For research use only; not for human or diagnostic use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.