Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GDC-0276 50mg | 1494581-70-2 | 50 MG
SDP

Catalog No. 5000229584
Encompass_Preferred

Selective, orally active NaV1.7 inhibitor research compound intended for preclinical and clinical pharmacology studies. Demonstrates sub-nanomolar potency and favorable pharmacokinetics; supplied as a high-purity small-molecule solid for laboratory use.

  • High purity (99.7%).
  • Molecular weight 462.58.
  • Chemical formula C24H31FN2O4S.
  • Reported potency IC50 = 0.4 nM (NaV1.7).
  • Supplied quantity 50 MG.

Catalog No. 50-002-29584 Supplier Medchemexpress LLC Supplier No. HY11423750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Selective, orally active NaV1.7 inhibitor research compound intended for preclinical and clinical pharmacology studies. Demonstrates sub-nanomolar potency and favorable pharmacokinetics; supplied as a high-purity small-molecule solid for laboratory use.

  • High purity (99.7%).
  • Molecular weight 462.58.
  • Chemical formula C24H31FN2O4S.
  • Reported potency IC50 = 0.4 nM (NaV1.7).
  • Supplied quantity 50 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.