Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GDC-0276 5mg | 1494581-70-2 | 5 MG
SDP

Catalog No. 5000227007
Encompass_Preferred

GDC-0276 is a potent, selective, reversible, orally active NaV1.7 inhibitor (IC50 = 0.4 nM) supplied as a white to off-white solid for laboratory research use.

  • Potent NaV1.7 inhibition (IC50 = 0.4 nM).
  • High purity: 99.69%.
  • Molecular weight: 462.58 g/mol; formula C24H31FN2O4S.
  • Soluble in DMSO (125 mg/mL); ultrasonic recommended.
  • Storage: powder at -20°C (3 years) or 4°C (2 years).
  • White to off-white solid appearance.

Catalog No. 50-002-27007 Supplier Medchemexpress LLC Supplier No. HY1142375MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GDC-0276 is a potent, selective, reversible, orally active NaV1.7 inhibitor (IC50 = 0.4 nM) supplied as a white to off-white solid for laboratory research use.

  • Potent NaV1.7 inhibition (IC50 = 0.4 nM).
  • High purity: 99.69%.
  • Molecular weight: 462.58 g/mol; formula C24H31FN2O4S.
  • Soluble in DMSO (125 mg/mL); ultrasonic recommended.
  • Storage: powder at -20°C (3 years) or 4°C (2 years).
  • White to off-white solid appearance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.