Learn More
Medchemexpress LLC GDC-0425 | 1200129-48-1 | 99.6% | 321.38 | 5 MG
SDP
Catalog No. 5000401712
Encompass_Preferred
GDC-0425 (RG-7602) is an orally available, highly selective small molecule ChK1 inhibitor. It can be used for the research of various malignancies.
  • Orally available
  • Highly selective small molecule ChK1 inhibitor
  • Can be used for the research of various malignancies
  • Exhibits partial suppression of tumor growth
  • Combination with Gemcitabine results in significant tumor regression in all tested models
Catalog No. 50-004-01712 Supplier Medchemexpress LLC Supplier No. HY199265MG
GDC-0425 (RG-7602) is an orally available, highly selective small molecule ChK1 inhibitor. It can be used for the research of various malignancies.
  • Orally available
  • Highly selective small molecule ChK1 inhibitor
  • Can be used for the research of various malignancies
  • Exhibits partial suppression of tumor growth
  • Combination with Gemcitabine results in significant tumor regression in all tested models