Learn More
Apexbio Technology LLC GDC-0449 (Vismodegib) 879085-55-9 200mg
SDP
Catalog No. 5000064218
Encompass_Preferred
GDC-0449 (Vismodegib CAS 879085-55-9) is a small molecule that selectively inhibits the Hedgehog (Hh) signaling pathway by binding to the Smoothened (SMO) receptor thus preventing downstream activation of Hh target genes implicated in cellular proliferation and differentiation This compound emerged from screening small-molecule libraries followed by medicinal chemistry optimization Preclinical evaluation demonstrated antitumor activity in murine models of medulloblastoma and xenograft models derived from human colon and pancreatic cancers Currently GDC-0449 is under investigation for locally advanced and metastatic solid tumors
Catalog No. 50-000-64218 Supplier Apexbio Technology LLC Supplier No. A3021200
GDC-0449 (Vismodegib CAS 879085-55-9) is a small molecule that selectively inhibits the Hedgehog (Hh) signaling pathway by binding to the Smoothened (SMO) receptor thus preventing downstream activation of Hh target genes implicated in cellular proliferation and differentiation This compound emerged from screening small-molecule libraries followed by medicinal chemistry optimization Preclinical evaluation demonstrated antitumor activity in murine models of medulloblastoma and xenograft models derived from human colon and pancreatic cancers Currently GDC-0449 is under investigation for locally advanced and metastatic solid tumors