Learn More
Apexbio Technology LLC GDC-0449 (Vismodegib), 500mg. Cas: 879085-55-9 MFCD: MFCD12407408. Hedgehog antagonist,potent and selective
SDP
Supplier:  Apexbio Technology LLC A3021500
Encompass_Preferred
GDC-0449 (2-chloro-N-[4-chloro-3-pyridin-2-yl-phenyl]-4-methane-sulfonyl benzamide), discovered by high throughput screening of a small molecule compound library followed by subsequent optimization through medical chemistry, is a potent and selective inhibitor of hedgehog (Hh) signaling. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4783