Learn More
Selleck Chemical LLC GDC-0575
SDP
Catalog No. 5000000867
Encompass_Preferred
GDC-0575 is a potent and selective CHK1 inhibitor with an IC50 of 1 2 nM
Catalog No. 50-000-00867 Supplier Selleck Chemical LLC Supplier No. S852625MG
GDC-0575 is a potent and selective CHK1 inhibitor with an IC50 of 1 2 nM