Learn More
Medchemexpress LLC HY-112167 5mg Medchemexpress, GDC-0575 CAS:1196541-47-5 Purity:>98%
SDP
Catalog No. 501969048
Encompass_Preferred
Medchemexpress, HY-112167 5mg GDC-0575 CAS:1196541-47-5 GDC-0575 (ARRY-575, RG7741) is a highly-selective oral small-molecule Chk1 inhibitor with an IC50 of 1.2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9048 Supplier Medchemexpress LLC Supplier No. HY1121675MG
Medchemexpress, HY-112167 5mg GDC-0575 CAS:1196541-47-5 GDC-0575 (ARRY-575, RG7741) is a highly-selective oral small-molecule Chk1 inhibitor with an IC50 of 1.2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.