Learn More
Medchemexpress LLC HY-112167A 5mg Medchemexpress, GDC-0575 dihydrochloride CAS:1657014-42-0 Purity:>98%
SDP
Catalog No. 501969049
Encompass_Preferred
Medchemexpress, HY-112167A 5mg GDC-0575 diHCl CAS:1657014-42-0 GDC-0575 dihydrochloride (ARRY-575 dihydrochloride) is an orally bioavailable CHK1 inhibitor, with an IC50 of 1.2 nM, and has antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9049 Supplier Medchemexpress LLC Supplier No. HY112167A5MG
Medchemexpress, HY-112167A 5mg GDC-0575 diHCl CAS:1657014-42-0 GDC-0575 dihydrochloride (ARRY-575 dihydrochloride) is an orally bioavailable CHK1 inhibitor, with an IC50 of 1.2 nM, and has antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.